Merck
  • Expression of CXCL1 in human endothelial cells induces angiogenesis through the CXCR2 receptor and the ERK1/2 and EGF pathways.
Expression of CXCL1 in human endothelial cells induces angiogenesis through the CXCR2 receptor and the ERK1/2 and EGF pathways.
Laboratory investigation; a journal of technical methods and pathology (2013-06-05)
Makito Miyake, Steve Goodison, Virginia Urquidi, Evan Gomes Giacoia, Charles J Rosser
ZUSAMMENFASSUNG
Endothelial cell growth and proliferation are critical for angiogenesis; thus, greater insight into the regulation of pathological angiogenesis is greatly needed. Previous studies have reported on chemokine (C-X-C motif) ligand 1 (CXCL1) expression in epithelial cells and that secretion of CXCL1 from these epithelial cells induces angiogenesis. However, limited reports have demonstrated CXCL1 expression in endothelial cells. In this report, we present data that expand on the role of CXCL1 in human endothelial cells inducing angiogenesis. Specifically, CXCL1 is expressed and secreted from human endothelial cells. Interference of CXCL1 function using neutralizing antibodies resulted in a reduction in endothelial cell migration and viability/proliferation, the latter associated with a decrease in levels of cyclin D and cdk4. In vitro studies revealed that CXCL1 influenced neoangiogenesis through the regulation of epidermal growth factor and ERK1/2. In a xenograft angiogenesis model, interference of CXCL1 function resulted in inhibition of angiogenesis. A better understanding of the role of CXCL1 in the interactions between the endothelial and epithelial components will provide insight into how human tissues use CXCL1 to survive and thrive in a hostile environment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Epidermaler Wachstumsfaktor aus Maussubmaxillardrüse, EGF, suitable for cell culture
Sigma-Aldrich
Drabkin-Reagenz, for the determination of hemoglobin
Sigma-Aldrich
Epidermaler Wachstumsfaktor aus Maussubmaxillardrüse, lyophilized powder, BioReagent, suitable for cell culture