Merck
  • Activation of type 2 cannabinoid receptors (CB2R) promotes fatty acid oxidation through the SIRT1/PGC-1α pathway.
Activation of type 2 cannabinoid receptors (CB2R) promotes fatty acid oxidation through the SIRT1/PGC-1α pathway.
Biochemical and biophysical research communications (2013-06-12)
Xuqin Zheng, Tao Sun, Xiaodong Wang
ZUSAMMENFASSUNG
Abnormal fatty acid oxidation has been associated with obesity and type 2 diabetes. At the transcriptional level, peroxisome proliferator-activated receptor-gamma coactivator 1α (PGC-1α) has been reported to strongly increase the ability of hormone nuclear receptors PPARα and ERRα to drive transcription of fatty acid oxidation enzymes. In this study, we report that a specific agonist of the type 2 cannabinoid receptor (CB2R) can lead to fatty acid oxidation through the PGC-1α pathway. We have found that CB2R is expressed in differentiated C2C12 myotubes, and that use of the specific agonist trans-caryophyllene (TC) stimulates sirtuin 1 (SIRT1) deacetylase activity by increasing the phosphorylation of cAMP response element-binding protein (CREB), thus leading to increased levels of PGC-1α deacetylation. This use of TC treatment increases the expression of genes linked to the fatty acid oxidation pathway in a SIRT1/PGC-1α-dependent mechanism and also drastically accelerates the rate of complete fatty acid oxidation in C2C12 myotubes, neither of which occur when CB2R mRNA is knocked down using siRNA. These results reveal that activation of CB2R by a selective agonist promotes lipid oxidation through a signaling/transcriptional pathway. Our findings imply that pharmacological manipulation of CB2R may provide therapeutic possibilities to treat metabolic diseases associated with lipid dysregulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-trans-Caryophyllen, ≥80%, FCC, FG
Sigma-Aldrich
Zyklisches Adenosin-3′,5′-Monophosphat, ≥98.5% (HPLC), powder
Sigma-Aldrich
Cyclisches Adenosin-3′,5′-monophosphat Natriumsalz Monohydrat, ≥98.0% (HPLC), powder
Sigma-Aldrich
(−)-trans-Caryophyllen, ≥98.0% (sum of enantiomers, GC)
Supelco
(−)-trans-Caryophyllen, analytical standard