- Replay: being sensitive: to specify when amino acid tracers accumulate in a brain lesion.
Replay: being sensitive: to specify when amino acid tracers accumulate in a brain lesion.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2013-06-13)
Karl-Josef Langen
PMID23757488
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Tyrosin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tyrosin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland