Merck
  • An alternative local anaesthesia technique to reduce pain in paediatric patients during needle insertion.
An alternative local anaesthesia technique to reduce pain in paediatric patients during needle insertion.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry (2013-06-14)
S H Lee, N Y Lee
ZUSAMMENFASSUNG
Pain control, which is necessary during most dental procedures, is administered by injecting a local anaesthetic. Because the injection itself can be painful, the procedure via which pain is reduced warrants continued investigation. Only a few studies regarding the reaction of children to dental needle insertion without the use of topical anaesthetics have been reported. This study was conducted to evaluate the efficacy of the local anaesthetic procedure without topical application as compared to the conventional insertion technique for alleviating pain in children receiving local anaesthesia injections. For the alternative injection procedure, the dentist quickly and gently pulled or pushed the clean and dried loose tissue at the injection site over the tip of the needle to a depth of 1 to 1.5 mm. When the end of the bevel of the needle tip entered the tissue, a few drops of solution were released, after which the needle was advanced to its proper and intended depth to continue anaesthetic release. There was a significant difference regarding the pain response between the alternative insertion technique (less painful) and the conventional one according to Sound, Eye, and Motor (SEM) scale ratings (P < 0.000). No significant difference was observed in the response between the maxilla and mandible, nor between boys and girls, between the conventional and alternative techniques. This alternative technique can reduce discomfort in paediatric dental patients and allow the clinician to administer a superficial local anaesthesia injection before the needle is advanced into deeper tissue. This technique is simple, quick, devoid of additional costs, and potentially more effective than the conventional needle insertion method.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(−)-Epinephrin
Supelco
Lidocainhydrochlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lidocain, powder
Supelco
Lidocain, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Lidocain -hydrochlorid Monohydrat, solid
Sigma-Aldrich
Lidocain, analytical standard
Adrenalin, European Pharmacopoeia (EP) Reference Standard
Lidocainhydrochlorid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prilocain -hydrochlorid, ≥98% (TLC)
Lidocain, European Pharmacopoeia (EP) Reference Standard