Merck
Adhesion and biofilm formation by oral streptococci on different commercial brackets.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry (2013-06-14)
C Passariello, P Gigola
ZUSAMMENFASSUNG
To compare early bacterial adhesion and biofilm formation in vitro by different oral streptococci on a variety of commercial brackets. Adhesion and biofilm formation in vitro of 6 Streptococcus spp. on 15 different commercial brackets, in standard culture medium and in human saliva were evaluated by the MTT reduction assay. Significant differences were evidenced in both early adhesion and biofilm formation among the studied brackets and between the two conditions of growth. Gold brackets resulted less prone to colonisation compared to composite brackets. The growth rates of the tested species on the different tested materials were significantly different. The adopted experimental plan, dissecting the two phases of plaque formation on different brackets in different conditions, showed that composite brackets are more susceptible to adhesion and colonisation by streptococci, while the remaining tested brackets did not show differences that could be clinically relevant. Data suggest that different personal behaviours affecting the oral environment could significantly affect colonisation of brackets by oral streptococci.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aluminiumoxid, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Aluminiumoxid, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminiumoxid, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminiumoxid, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminiumoxid, Corundum, α-phase, -100 mesh
Supelco
Aluminiumoxid, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminiumoxid, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminiumoxid, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminiumoxid, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminiumoxid, 99.997% trace metals basis
Sigma-Aldrich
Aluminiumoxid, single crystal substrate, <0001>
Sigma-Aldrich
Aluminiumoxid, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminumoxid, mesoporös, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminiumoxid, for the determination of hydrocarbons