- How to interpret thyroid function tests.
How to interpret thyroid function tests.
Clinical medicine (London, England) (2013-06-14)
Olympia Koulouri, Mark Gurnell
PMID23760704
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thyreotropes Hormon aus human pituitary, lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)
Supelco
3,3′,5-Triiodo-L-thyronin (T3) -Lösung, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
L-Thyroxin (T4) -Lösung, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®