Merck
  • Effect of high LH/FSH ratio on women with polycystic ovary syndrome undergoing in vitro maturation treatment.
Effect of high LH/FSH ratio on women with polycystic ovary syndrome undergoing in vitro maturation treatment.
The Journal of reproductive medicine (2013-06-15)
Amir Wiser, Fady Shehata, Hananel Holzer, Jordana H Hyman, Einat Shalom-Paz, Weon-Young Son, Togas Tulandi
ZUSAMMENFASSUNG
To determine whether high luteinizing hormone/follicle-stimulating hormone (LH/FSH) ratios have a clinical impact on women with polycystic ovary syndrome (PCOS) undergoing in vitro maturation (IVM) treatment. Women with PCOS who underwent IVM treatment were divided into those with LH/FSH ratio > 1.5 and LH/FSH 0.5-1.5. We analyzed baseline characteristics of the patients, number of oocytes retrieved, number of mature oocytes, and pregnancy rates. Women with LH/FSH ratio of > 1.5 had higher basal serum testosterone (2.2 vs. 1.4, p < 0.005, CI 0.1-1.0) and estradiol (188.7 +/- 16.2 vs. 143.7 +/- 6.9, p < 0.01, CI 23-96). The antral follicle count (AFC) was also higher in the patients with high LH/FSH (46.2 +/- 3.5 vs. 32.9 +/- 1.3, p < 0.001, CI 7-21). The total number of retrieved oocytes and number of mature oocytes was also significantly higher in women with LH/FSH ratio of > 1.5 than in those with a lower ratio. However, the pregnancy rate in women with LH/FSH ratio of > 1.5 (16.7%) was significantly lower than in those with a ratio of 0.5-1.5 (40.4%), p < 0.05, odds ratio 0.32. PCOS patients with LH/FSH ratio of > 1.5 had higher basal testosterone, E2, and AFC but decreased pregnancy rate. This could be due to the deleterious effect of LH on folliculogenesis and endometrial receptivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Follikelstimulierendes Hormon aus human pituitary, ~7,000 IU/mg (powder)
Sigma-Aldrich
Testosteron, purum, ≥99.0% (HPLC)
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
17β-Estradiol -Lösung, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Estradiol, analytical standard
Supelco
Testosteron, VETRANAL®, analytical standard
Testosteron, European Pharmacopoeia (EP) Reference Standard
Estradiol Hemihydrat, European Pharmacopoeia (EP) Reference Standard