Merck
Clinical experience of integrative cancer immunotherapy with GcMAF.
Anticancer research (2013-06-20)
Toshio Inui, Daisuke Kuchiike, Kentaro Kubo, Martin Mette, Yoshihiro Uto, Hitoshi Hori, Norihiro Sakamoto
ZUSAMMENFASSUNG
Immunotherapy has become an attractive new strategy in the treatment of cancer. The laboratory and clinical study of cancer immunotherapy is rapidly advancing. However, in the clinical setting, the results of cancer immunotherapy are mixed. We therefore contend that cancer immunotherapy should be customized to each patient individually based on their immune status and propose an integrative immunotherapy approach with second-generation group-specific component macrophage activating factor (GcMAF)-containing human serum. The standard protocol of our integrative cancer immunotherapy is as follows: i) 0.5 ml GcMAF-containing human serum is administered intramuscularly or subcutaneously once or twice per week for the duration of cancer therapy until all cancer cells are eradicated; ii) hyper T/natural killer (NK) cell therapy is given once per week for six weeks; iii) high-dose vitamin C is administered intravenously twice per week; iv) alpha lipoic acid (600 mg) is administered orally daily; v) vitamin D3 (5,000-10,000 IU) is administered orally daily. By March 2013, Saisei Mirai have treated over 345 patients with GcMAF. Among them we here present the cases of three patients for whom our integrative immunotherapy was remarkably effective. The results of our integrative immunotherapy seem hopeful. We also plan to conduct a comparative clinical study.>
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
(+)-Natrium-L-ascorbat, crystalline, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(+)-Natrium-L-ascorbat, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Natrium-L-ascorbat, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Supelco
Natriumascorbat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbinsäure, tested according to Ph. Eur.