Merck
  • Polyethylene wear and osteolysis after cementless total hip arthroplasty with alumina-on-highly cross-linked polyethylene bearings in patients younger than thirty years of age.
Polyethylene wear and osteolysis after cementless total hip arthroplasty with alumina-on-highly cross-linked polyethylene bearings in patients younger than thirty years of age.
The Journal of bone and joint surgery. American volume (2013-06-21)
Young-Hoo Kim, Jang-Won Park, Chirag Patel, Dae-Youn Kim
ZUSAMMENFASSUNG
We asked whether cementless total hip arthroplasties that made use of alumina-on-highly cross-linked polyethylene bearings would improve hip score and functional activity and reduce the prevalence of polyethylene wear, osteolysis, and aseptic loosening. Consecutive primary total hip arthroplasties were performed in fifty patients (a total of sixty hips among thirty-four men and sixteen women) who were younger than thirty years of age. The average age at the time of the index arthroplasty was 28.3 years (range, twenty-one to twenty-nine years). The average follow-up was 10.8 years (range, ten to twelve years). Osteolysis and polyethylene wear rates were evaluated with use of radiography and computed tomography. The mean Harris hip score, which was 38 points (range, 6 to 45 points) preoperatively, had improved to 95 points (range, 85 to 100 points) at a mean follow-up time of 10.8 years. The mean penetration (and standard error of the mean) of the polyethylene liner was 0.031 ± 0.004 mm per year. No hip had osteolysis or aseptic loosening. At a minimum of ten years and an average of 10.8 years postoperatively, the current generation of cementless acetabular and femoral components with alumina-on-highly cross-linked polyethylene bearings was functioning well and was not associated with the development of osteolysis in our group of patients younger than thirty years of age. While the long-term prevalence of polyethylene wear and osteolysis remains unknown, the midterm data are promising.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aluminiumoxid, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminiumoxid, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminiumoxid, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminiumoxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminiumoxid, powder, 99.99% trace metals basis
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylen, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Aluminiumoxid, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Polyethylen, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Aluminiumoxid, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Aluminiumoxid, Corundum, α-phase, -100 mesh
Supelco
Aluminiumoxid, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminiumoxid, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminiumoxid, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Polyethylen, High density, melt index 12 g/10 min (190 °C/2.16kg)
Polyethylen (LDPE), ERM®, certified reference material
Sigma-Aldrich
Aluminiumoxid, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Polyethylen, Medium density
Sigma-Aldrich
Aluminiumoxid, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminiumoxid, 99.997% trace metals basis
Sigma-Aldrich
Polyethylen, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylen, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylen, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Aluminiumoxid, single crystal substrate, <0001>
Sigma-Aldrich
Aluminiumoxid, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminumoxid, mesoporös, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Polyethylen, analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Supelco
Aluminiumoxid, for the determination of hydrocarbons