- Corticosteroids can reduce the severity of scoliosis in Duchenne muscular dystrophy.
Corticosteroids can reduce the severity of scoliosis in Duchenne muscular dystrophy.
The Journal of bone and joint surgery. American volume (2013-06-21)
Anne M Connolly, Han Jo Kim, Keith H Bridwell
PMID23783217
MATERIALIEN