Merck
  • Astrocytes use a novel transporter to fill gliotransmitter vesicles with D-serine: evidence for vesicular synergy.
Astrocytes use a novel transporter to fill gliotransmitter vesicles with D-serine: evidence for vesicular synergy.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-06-21)
Mari Sild, Marion R Van Horn
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Serin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Serin, BioUltra, ≥99.5% (NT)
Supelco
L-Serin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland