Merck
  • Repellency of cassia bark, eucalyptus, and star anise oils and their major constituents to Leptotrombidium pallidum (Acari: Trombiculidae).
Repellency of cassia bark, eucalyptus, and star anise oils and their major constituents to Leptotrombidium pallidum (Acari: Trombiculidae).
Journal of medical entomology (2013-06-28)
E-Hyun Shin, Bong Gu Song, Il Hee Lee, Mi Yeoun Park, Young-Joon Ahn, Kyu-Sik Chang
ZUSAMMENFASSUNG
Leptotrombidium pallidum (Nagoya, Miyagawa, Mitamura & Tamiya) is a primary vector of Orientia tsutsugamushi (Hyashi), the causative agent of scrub typhus. An assessment is made of the repellency to L. pallidum larvae (chiggers) of cassia bark, eucalyptus, and star anise oils and major constituents (E)-cinnamaldehyde, 1,8-cineole, and (E)-anethole of the corresponding oils. Results were compared with those of conventional repellents DEET (N,N-diethyl-3-methylbenzamide), IR3535 [(ethyl 3-[acetyl(butyl)amino]propanoate)], and permethrin. Based on the median repellent concentration (RC50) values, (E)-cinnamaldehyde, (E)-anethole, cassia bark oil, and star anise oil (RC50, 0.95-1.52 mg/cm2) exhibited significantly more potent repellency than DEET (3.85 mg/cm2). (E)-cinnamaldehyde, (E)-anethole, cassiabark oil, 1,8-cineole, and star anise oil were approximately 43, 16, 11, 8, and 4 times more effective than IR3535 (CC5, 6.51%) as judged by the median climbing distance-disturbing concentration (CC50) values. The median residual duration time of repellency (RT50) was significantly more pronounced in DEET (RT50, 323 min) than in all essential oils and constituents (108-167 min). In the light of global efforts to reduce the level of highly toxic synthetic repellents, the three essential oils and their major constituents described merit further study as potential biorepellents for the control of L. pallidum populations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eucalyptol, 99%
Sigma-Aldrich
Zimtaldehyd, natural, ≥95%, FG
Sigma-Aldrich
trans--Zimtaldehyd, 97%
Sigma-Aldrich
trans-Anethol, 99%
Sigma-Aldrich
trans--Zimtaldehyd, ≥99%
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Sigma-Aldrich
trans--Zimtaldehyd, FCC, FG
Supelco
Eucalyptol, analytical standard
Sigma-Aldrich
trans-Anethol, ≥99%, FCC, FG
Supelco
1,8-Cineol, primary reference standard
Supelco
trans-Anethol, analytical standard
Supelco
trans-Anethol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
trans--Zimtaldehyd, analytical standard
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.
Cineol, European Pharmacopoeia (EP) Reference Standard