Merck
  • Fasting insulin serum levels and psychopathology profiles in male schizophrenic inpatients treated with olanzapine or risperidone.
Fasting insulin serum levels and psychopathology profiles in male schizophrenic inpatients treated with olanzapine or risperidone.
Neuro endocrinology letters (2013-06-28)
Beata Konarzewska, Napoleon Waszkiewicz, Beata Galińska, Agata Szulc
ZUSAMMENFASSUNG
Recent studies have suggested that higher insulin levels are associated with better psychopathology profiles in cross-sectional samples of patients with schizophrenia. This study examines whether drug-induced fasting insulin changes between third and eight week of treatment are related to clinical improvement in non-diabetic patients receiving the atypical neuroleptics: risperidone or olanzapine. non-diabetic men with a diagnosis of schizophrenia according to the DSM-IV diagnostic classification were recruited from psychiatric inpatient units. Following a drug-free period, neuroleptic treatment was initiated (risperidone n=36, olanzapine n=35) and doses were adjusted to achieve maximal clinical efficacy. All patients were hospitalized throughout the study. Initial and final evaluations of serum insulin levels and psychopathology (assessed with the Positive and Negative Syndrome Scale, PANSS), were carried out at weeks 3 and 8 after the onset of treatment, respectively. There were no differences between and within the risperidone and olanzapine groups in changes of serum insulin level between the third and eighth week of treatment. In the olanzapine group, Pearson correlation analysis revealed a significant negative correlation between changes in fasting serum insulin levels and the PANSS-Total, Positive and General Psychopathology subscale scores. Only improvement in the PANSS-Negative Symptom subscale score was not correlated with insulin level change between the third and eighth week of treatment. In the risperidone group, correlations between PANSS subscales scores and the corresponding serum insulin levels change were positive, albeit statistically non-significant. In both groups the improvement in PANSS-Total scores was not correlated with changes in BMI. Olanzapine-related changes in endogenous fasting insulin levels were correlated with clinical improvement in acutely ill non-diabetic schizophrenic patients. Because the interesting linkage between insulin and positive and negative symptoms could be an epiphenomenon, randomized studies are needed to further explore the role of insulin in therapeutic responses in patients with schizophrenia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Risperidon, ≥98% (HPLC), powder
Supelco
Olanzapine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Olanzapin, ≥98% (HPLC)
Supelco
Risperidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Risperidon für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Risperidon, European Pharmacopoeia (EP) Reference Standard