Merck
  • NG2 regulates directional migration of oligodendrocyte precursor cells via Rho GTPases and polarity complex proteins.
NG2 regulates directional migration of oligodendrocyte precursor cells via Rho GTPases and polarity complex proteins.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-06-28)
Fabien Binamé, Dominik Sakry, Leda Dimou, Valérie Jolivel, Jacqueline Trotter
ZUSAMMENFASSUNG
The transmembrane proteoglycan NG2 is expressed by oligodendrocyte precursor cells (OPC), which migrate to axons during developmental myelination and remyelinate in the adult after migration to injured sites. Highly invasive glial tumors also express NG2. Despite the fact that NG2 has been implicated in control of OPC migration, its mode of action remains unknown. Here, we show in vitro and in vivo that NG2 controls migration of OPC through the regulation of cell polarity. In stab wounds in adult mice we show that NG2 controls orientation of OPC toward the wound. NG2 stimulates RhoA activity at the cell periphery via the MUPP1/Syx1 signaling pathway, which favors the bipolar shape of migrating OPC and thus directional migration. Upon phosphorylation of Thr-2256, downstream signaling of NG2 switches from RhoA to Rac stimulation. This triggers process outgrowth through regulators of front-rear polarity and we show using a phospho-mimetic form of NG2 that indeed NG2 recruits proteins of the CRB and the PAR polarity complexes to stimulate Rac activity via the GEF Tiam1. Our findings demonstrate that NG2 is a core organizer of Rho GTPase activity and localization in the cell, which controls OPC polarity and directional migration. This work also reveals CRB and PAR polarity complexes as new effectors of NG2 signaling in the establishment of front-rear polarity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ribonukleinsäure aus Torulahefe, Type VI
SAFC
L-Threonin
Sigma-Aldrich
Ribonukleinsäure aus Backhefe (S. cerevisiae)
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
Supelco
L-Threonin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ribonukleinsäure aus Torulahefe, Type IX
Supelco
L-Threonin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ribonukleinsäure aus Torulahefe, core, Type II-C