Merck
[Nutrition value of tropical and subtropical fruits].
Voprosy pitaniia (2013-07-03)
G G Dubtsov, V V Bessonov, V G Baĭkov, N N Makhova, L V Sheviakova, M N Bogachuk, E K Baĭgarin, Lazar Iao Bru
ZUSAMMENFASSUNG
The article is devoted to the study of the chemical composition of tropical and subtropical fruit (avocado, papaya and mango), which are now in great numbers are on the appeared on the Russian market. Due to use technology tropical and subtropical fruits can be implemented in almost all areas and regions of the country. Relatively low cost makes these products quite popular among the people. In domestic scientific literature there are no systematic data describing the chemical composition of these tropical and subtropical fruits sold in the domestic market, while the information needed to calculate food and energy value of diets and culinary products derived from tropical and subtropical fruit. Avocado fruits are sources of insoluble dietary fiber content of which was equal to 12.2%, as well as minerals. The study of the fatty acid composition of lipids avocados showed high content of oleic acid fruit, which accounts for 53.2% of total fatty acids in these fruits. Which makes them a valuable source of unsaturated fatty acids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Supelco
Oleinsäure, analytical standard
Supelco
Oleinsäure, Selectophore, ≥99%