- Post annealing effects on the electrical characteristics of pentacene thin film transistors on flexible substrates.
Post annealing effects on the electrical characteristics of pentacene thin film transistors on flexible substrates.
Journal of nanoscience and nanotechnology (2013-07-19)
Tae-Yeon Oh, Shin Woo Jeong, Seongpil Chang, Jung-Ho Park, Jong-Woo Kim, Kookhyun Choi, Hyeon-Jun Ha, Bo-Yeon Hwang, Byeong-Kwon Ju
PMID23858886
ZUSAMMENFASSUNG
This work studies the effect of post annealing of pentacene on a flexible substrate through the examination of electrical properties and surface morphologies. It is confirmed that the best performance of devices is achieved when the post annealing temperature is 60 degrees C, since the grain size increases, which decrease grain boundaries caused charge transport limit. We can also confirmed the large threshold voltage shift of device annealed at 60 degrees C that means the lower trap density between channel and insulator interface. The device annealed at 60 degrees C exhibits a saturation mobility of 1.99 cm2/V x s, an on/off ratio of 1.87 x 10(4), and a subthreshold slope of 2.5 V/decade.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Titan(IV)-oxid, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Rutil, nanopowder, <100 nm particle size, 99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Mischung aus Rutil und Anatas, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, Mischung aus Rutil und Anatas, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titan(IV)-oxid, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titan(IV)-oxid, Rutil, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm