Merck
Water-soluble and biocompatible MnO@PVP nanoparticles for MR imaging in vitro and in vivo.
Journal of biomedical nanotechnology (2013-07-19)
Xiaoqing Hu, Yuxuan Ji, Mingliang Wang, Fei Miao, Hongmei Ma, Hebai Shen, Nengqin Jia
ZUSAMMENFASSUNG
The uniform-sized manganese oxide nanoparticles (the oleic-capped MnO NPs) were synthesized by the thermal decomposition of Mn-oleate complex and were transferred into water with the help of cationic surfactant of cetyltrimethyl ammonium bromide (CTAB), then the poly(vinylpyrrolidone) (PVP) membrane was further coated on to them with the aid of anionic dispersant of poly(styrenesulfonate) (PSS) by layer-by-layer electrostatic assembly to render them water soluble and biocompatible. They were characterized by transmission electron microscopy (TEM), dynamic light scattering (DLS), X-ray diffraction (XRD), Fourier-transform infrared (FTIR) and MTT assay. In vitro cellular uptake test revealed the MnO@PVP NPs were low cytotoxic, biocompatible and could be used as a T,-positive contrast agent for passive targeting magnetic resonance imaging (MRI). Interestingly, signal enhancement in cerebral spinal fluid (CSF) spaces in vivo experiment suggested that the MnO@PVP NPs can pass through the blood brain barrier (BBB). These results show that MnO@PVP NPs are good candidates as MRI contrast agents with the lack of cytotoxicity and have great potential applications in magnetic nano-device and biomagnetic field.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polyvinylpyrrolidon, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidon, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
Polyvinylpyrrolidon, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidon, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidon, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidon, K 30
Sigma-Aldrich
Polyvinylpyrrolidon, K 90
Sigma-Aldrich
Polyvinylpyrrolidon, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidon, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
Polyvinylpyrrolidon -Lösung, K 60, 45% in H2O
Sigma-Aldrich
Polyvinylpyrrolidon, Molecular Biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Mangan(II,III)-oxid, 97%
Supelco
Povidon, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Polyvinylpyrrolidon, K 25, tested according to Ph. Eur.
Sigma-Aldrich
Polyvinylpyrrolidon, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Povidon, meets USP testing specifications
Povidon, European Pharmacopoeia (EP) Reference Standard