- [Features of spatial orientation formation in posterity of female rats with liver experimental defeat in Morris water maze].
[Features of spatial orientation formation in posterity of female rats with liver experimental defeat in Morris water maze].
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova (2013-07-19)
V A Vakhnin, G V Brukhin
PMID23862386
ZUSAMMENFASSUNG
It was established, that the chronic experimental hepatobiliary systems alcohol induced pathology of female rats caused infringement of spatial differentiation development in their posterity in Morris water maze. In experimental infant rats it was revealed the increase of time of platform fin ding in a water Morris task, lengths of trajectory of movement, increase of time of stay near walls (thigmotaxis) and speed of movement reduction, the latent period of zones of start leaving and efficiency of way index.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 Ampullen/Kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 Ampullen/Kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 Ampullen/Kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol -Lösung, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®