Merck
  • Evaluation of calcium, magnesium, potassium and sodium in biological samples of male human immunodeficiency virus patients with tuberculosis and diarrhea compared to healthy control subjects in Pakistan.
Evaluation of calcium, magnesium, potassium and sodium in biological samples of male human immunodeficiency virus patients with tuberculosis and diarrhea compared to healthy control subjects in Pakistan.
Clinical laboratory (2013-07-20)
Hassan Imran Afridi, Tasneem Gul Kazi, Farah Naz Talpur, Naveed Kazi, Faheem Shah Naeemullah, Sadaf Sadia Arain, Kapil Dev Brahman
ZUSAMMENFASSUNG
Electrolyte deficiency has been associated with an increased risk of human immunodeficiency virus type 1 (HIV-1) disease progression and mortality. This study examined the association between low electrolyte concentrations in blood and scalp hair and the presence of opportunistic infections in patients with acquired immune deficiency syndrome (AIDS). Sixty-two male HIV positive patients (HIV-1) from various cities in Pakistan were recruited to the study. These Patients were divided into two groups according to secondary infections (tuberculosis and high fever with diarrhea), and biological samples (scalp hair, serum, blood and urine) were collected from them. As a comparative control group, 120 healthy subjects (males) of the same age group (31 - 45 years), socio-economic status, localities and dietary habits were also included in the study. The elements in the biological samples were analyzed by flame atomic absorption spectrophotometry after microwave-assisted acid digestion. Validity and accuracy of the methodology were checked using certified reference materials (CRMs) and against values obtained by a conventional wet acid digestion method on the same CRMs. The results indicated significantly lower levels of calcium, potassium, magnesium and natrium in all analyzed biological samples (blood, serum and scalp hair) of male patients with Acquired Immune Deficiency Syndrome (AIDS) in comparison to healthy controls (p < 0.01), while the levels of these elements were found to be higher in urine samples of the AIDS patients than in those of the control group. These data offer guidance to clinicians and other professionals investigating the deficiency of electrolytes in biological samples (scalp hair, serum and blood) of AIDS patients in relation to healthy subjects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natrium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Natrium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
Natrium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Natrium, ACS reagent, dry
Sigma-Aldrich
Natrium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium Zubereitung, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium (in Sure/Seal Flasche), in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium (in Sure/Seal Flasche), in a Sure/Seal bottle, turnings, 37.5 mmol