Merck
  • Hydrogen-bonding linkage of thymidine derivatives with carboxylic acid and pyridyl groups in a crystalline state.
Hydrogen-bonding linkage of thymidine derivatives with carboxylic acid and pyridyl groups in a crystalline state.
Journal of nanoscience and nanotechnology (2013-08-02)
Junichi Hoshino, Junpei Kuwabara, Takaki Kanbara
ZUSAMMENFASSUNG
Thymidine derivatives with carboxylic acid and pyridyl groups were synthesized for constructing one-dimensional network structure based on hydrogen bonding in crystalline state. The solid sate structures and hydrogen bonding networks of the thymidine derivatives were characterized by single X-ray diffraction analysis. The thymidine derivatives formed a zwitterion structure with a pyridinium proton and a carboxylate moiety in a crystalline state due to transfer of a proton from the carboxylic acid to the pyridyl moiety. Strong hydrogen bonds between the pyridinium proton and the carboxylate moiety connected the thymidine units, resulting in a one-dimensional polymeric structure with a uniform direction reminiscent of the structure of single-strand polythymidine. The chemical structure of the pyridyl group affects the hydrogen-bonding networks. The well-designed hydrogen-bonding interaction served as connecting parts for polythymidine mimics even in the presence of other hydrogen-bonding motifs such as nucleobases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidin, ≥99%
Sigma-Aldrich
Pyridin -hydrochlorid, 98%
Sigma-Aldrich
Pyridin, ≥99%
Sigma-Aldrich
Pyridin, biotech. grade, ≥99.9%
Sigma-Aldrich
Thymidin, ≥99.0% (HPLC)
Supelco
Pyridin, analytical standard
Sigma-Aldrich
Pyridin -hydrobromid, 98%
Sigma-Aldrich
Pyridin -hydrochlorid, purum, ≥98.0% (AT)
Supelco
Pyridin -Lösung, certified reference material, 2000 μg/mL in methanol