Merck
  • Catalytic activity and structure properties of doped VOHPO4 x 0.5H2O with nanosized Ru, Au, Fe and Mn in benzene hydroxylation.
Catalytic activity and structure properties of doped VOHPO4 x 0.5H2O with nanosized Ru, Au, Fe and Mn in benzene hydroxylation.
Journal of nanoscience and nanotechnology (2013-08-02)
Peter R Makgwane, Suprakas Sinha Ray
ZUSAMMENFASSUNG
The promotional effect of nanosized Ru, Fe, Au, and Mn particles on VOHPO4 x 0.5H2O (VHP) catalytic properties was investigated in benzene hydroxylation reaction using hydrogen hydroperoxide (H2O2) as oxidant. Catalytic results indicated a profound effect of the nanoparticle dopants on VHP catalyst activity and products distribution. Amongst the promoted VHP catalysts, Au/VHP exhibited high catalytic effect with benzene conversion of 76% at a combined 85.5% selectivity toward the formation of phenol and hydroquinone achieved in 6 h under optimised reaction conditions. The extended scope application of nanosized doped Au-VHP showed to provide an effective catalyst for activation of the aromatic hydrocarbons C-H bonds into oxygenate derivatives. The catalyst could be re-used for several cycles with insignificant loss of activity. The doped nanosized Au-VHP catalyst provide a clean promising catalytic route based on heterogeneous catalysis for transformation of aromatics into value-added oxygenates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Supelco
Benzol, analytical standard
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Supelco
Benzol -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis