- New CAD/CAM materials and blocks for chairside procedures.
New CAD/CAM materials and blocks for chairside procedures.
International journal of computerized dentistry (2013-08-13)
M Zimmermann, A Mehl, S Reich
PMID23930578
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zirkonium(IV)-oxid -Lösung, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirkonium(IV)-oxid -Lösung, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O