Merck
  • Complete staining of human spermatozoa and immature germ cells combined with phase contrast microscopy.
Complete staining of human spermatozoa and immature germ cells combined with phase contrast microscopy.
Archives of andrology (1987-01-01)
A Y Michael, J O Drejer, P V Bagger, G U Detlefsen, G Stakemann
ZUSAMMENFASSUNG
A method combining Janus green B and Thymol blue stains the anterior part of the head, the nuclear membrane, middle piece, and tail of spermatozoa light green and the nucleus deep purple. The method provides excellent stained preparations for the evaluation of sperm morphology by phase contrast microscopy. It produces significantly less abnormal spermatozoa compared with the Papanicolaou stain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thymolblau, ACS reagent
Sigma-Aldrich
Janusgrün B, certified by the BSC, Dye content 65 %
Supelco
Thymolphthalein, ACS reagent
Sigma-Aldrich
Thymolphthalein, ACS reagent, Dye content 95 %
Sigma-Aldrich
Thymolblau, ACS reagent, Dye content 95 %