Merck
  • Single-step purification of pertussis toxin and its subunits by heat-treated fetuin-sepharose affinity chromatography.
Single-step purification of pertussis toxin and its subunits by heat-treated fetuin-sepharose affinity chromatography.
Biochemistry and cell biology = Biochimie et biologie cellulaire (1989-07-01)
P Chong, M Klein
ZUSAMMENFASSUNG
A general procedure for purifying biologically active pertussis toxin from Bordetella pertussis fermentation broth using affinity chromatography on heat-treated fetuin-Sepharose CL-4B is described. Diethanolamine is used as eluent in this single-step purification to prepare endotoxin-free pertussis toxin in good yield (70%) and high purity (greater than 95%). This one-step affinity chromatography procedure can be easily applied for large-scale preparation of pertussis toxin S1 subunit and its B-component. The affinity-purified S1 subunit is devoid of any of the histamine-sensitizing activity normally associated with pertussis toxin. The chromatographically purified pertussis toxin and its subunits retained their immunogenicity and could induce high levels of anti-toxin neutralizing antibodies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Sepharose CL-4B, Cross-linked