Merck
  • Allergic contact sensitization to the glycidyl ester of hexahydrophthalic acid in a cutting oil.
Allergic contact sensitization to the glycidyl ester of hexahydrophthalic acid in a cutting oil.
Contact dermatitis (1986-08-01)
J S English, I Foulds, I R White, R J Rycroft
ZUSAMMENFASSUNG
Allergic contact dermatitis is reported in 5 machine operatives working with a cutting oil containing an epoxy compound used as a stabilizer. Positive patch test reactions to this compound were obtained in all 5 men and were negative in 25 controls.