Merck
  • Determination of optical purity by high-performance liquid chromatography of compounds of pharmaceutical interest.
Determination of optical purity by high-performance liquid chromatography of compounds of pharmaceutical interest.
Il Farmaco; edizione pratica (1988-11-01)
G Gamberini, V Ferioli
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Amino-1-butanol, 97%