- Cellular and molecular pharmacology of auranofin and related gold complexes.
Cellular and molecular pharmacology of auranofin and related gold complexes.
Biochemical pharmacology (1986-10-15)
S T Crooke, R M Snyder, T R Butt, D J Ecker, H S Allaudeen, B Monia, C K Mirabelli
PMID3533080
MATERIALIEN