Merck
Arginyl residues and thermal stability in proteins.
Molecular and cellular biochemistry (1986-08-01)
F S Qaw, J M Brewer
ZUSAMMENFASSUNG
Guanidination and amidination of bovine serum albumin, yeast enolase and yeast alcohol dehydrogenase were accompanied by increases in thermal stability at lower extents of modification. Decreases in thermal stability result from greater modification. These results support suggestions that surface guanidino groups (arginyl groups) are an important factor in thermal stability of proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
O-Methyl-isoharnstoff -hemisulfat (Salz), 99%
Sigma-Aldrich
Methylacetimidat -hydrochlorid, technical grade
Sigma-Aldrich
o-Methylisoharnstoffbisulfat, 99%