Merck
Toxicity of n-hexane and 2,5-hexanediol to the haemopoietic system: a preliminary report.
Xenobiotica; the fate of foreign compounds in biological systems (1987-02-01)
S K Goel, G S Rao, C R Krishnamurti
ZUSAMMENFASSUNG
The effect of n-hexane and its metabolite, 2,5-hexanediol, on the haemopoietic system has been investigated in the rat. 59Fe uptake was decreased in the bone marrow of n-hexane-treated animals. Spleen and bone marrow were the target sites for the metabolite 2,5-hexanediol, whereas n-hexane as such does not affect these organs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,5-Hexandiol, 99% (mixture of isomers)