Merck
  • Topographical relationships between the monovalent and divalent cation binding sites of des-1-41-light chain bovine plasma protein C and des-1-41-light chain-activated bovine plasma protein C.
Topographical relationships between the monovalent and divalent cation binding sites of des-1-41-light chain bovine plasma protein C and des-1-41-light chain-activated bovine plasma protein C.
The Journal of biological chemistry (1987-05-25)
K A Hill, F J Castellino
ZUSAMMENFASSUNG
The paramagnetic effect of Mn2+ on the longitudinal relaxation rate (T1)-1 of 205Tl+, when both cations are bound to des-1-41-light chain bovine plasma protein C (GDPC) and its activation product, des-1-41-light chain-activated bovine plasma protein C (GDAPC), has been assessed by 205Tl+ NMR spectroscopy. A substantial shortening of the T1 for Tl+ bound to either protein was observed in the presence of Mn2+, an effect not noted upon substitution of Mn2+ with the diamagnetic cation Ca2+, which is known to bind to these proteins in a similar fashion to Mn2+. This paramagnetic effect was employed to estimate distances between the monovalent and divalent cation sites in these proteins, approximately 6.7 +/- 0.2 A with GDPC and 8.3 +/- 0.2 A in GDAPC. These data suggest that a conformational alteration occurs upon activation of GDPC which leads to an increase in the distance between the monovalent and divalent cation sites.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Protein C from human plasma, Activated, lyophilized powder, ≥90% (SDS-PAGE)