- Metabolism of 2,4-dinitrotoluene, 2,4-dinitrobenzyl alcohol and 2,4-dinitrobenzaldehyde by rat liver microsomal and cytosol fractions.
Metabolism of 2,4-dinitrotoluene, 2,4-dinitrobenzyl alcohol and 2,4-dinitrobenzaldehyde by rat liver microsomal and cytosol fractions.
Chemical & pharmaceutical bulletin (1987-04-01)
M Shoji, M Mori, T Kawajiri, M Sayama, Y Mori, T Miyahara, T Honda, H Kozuka
PMID3652297
MATERIALIEN