Merck
Metabolism of selenocyanate in the rat.
Archives of biochemistry and biophysics (1987-10-01)
S Vadhanavikit, R J Kraus, H E Ganther
ZUSAMMENFASSUNG
Rats injected subcutaneously with 2 mg Se/kg body weight of [75Se]selenocyanate or [14C, 75Se]selenocyanate excreted dimethylselenide (DMSe) in the breath and trimethyl-selenonium ion (TMSe) in the urine. The 24-h respiratory DMSe and urinary TMSe excretions were 26.8 +/- 8.1 and 14.5 +/- 5.1% of the dose, respectively. Tissue concentrations of 75Se were highest in the kidneys (1.89 +/- 0.2% dose/g), liver (1.46 +/- 0.2% dose/g), and blood (0.50 +/- 0.05% dose/ml), and lower (greater than 0.3% dose/g) in the other tissues. Trimethyl-selenonium was the major form (61%) of selenium in urine. Approximately 2% of the dose of doubly labeled SeCN- was excreted unchanged in urine (about 12% of urinary Se). 14C from doubly labeled SeCN- was not present in the methylated selenium metabolites, but a major 14C urinary metabolite was identified as thiocyanate. These results indicate that a substantial part of selenocyanate in the body undergoes metabolism and Se is excreted in methylated forms following scission of the C-Se bond.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumselencyanat, reagent grade, 97%
Sigma-Aldrich
Kaliumselencyanat, ReagentPlus®, ≥99%