Merck
Contact urticaria from diethyl fumarate.
Contact dermatitis (1985-03-01)
A Lahti, H I Maibach
ZUSAMMENFASSUNG
The contact urticariagenic properties of diethyl fumarate were studied using the guinea pig ear swelling assay and by open application on the human upper back skin. Diethyl fumarate caused non-immunologic contact urticaria in both human and guinea pig skin, and the reactions exhibited similar dose dependency and timing of maximal response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fumarsäure-diethylester, 98%