Merck
Evaluation of controlled release formulations of mosquito larvicides.
The Indian journal of medical research (1984-12-01)
M Kalyanasundaram, C M Reddy, T Mariappan, P K Das
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phthalsäure-di-n-octylester, ≥98.0% (GC)
Supelco
Phthalsäure-di-n-octylester, analytical standard
Supelco
Phthalsäure-di-n-octylester, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland