- Electron microscopic immunoperoxidase staining of insulin using 4-chloro-1-naphthol after osmium fixation.
Electron microscopic immunoperoxidase staining of insulin using 4-chloro-1-naphthol after osmium fixation.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1982-07-01)
D G Baskin, H Mar, K C Gorray, W Y Fujimoto
PMID6179987
ZUSAMMENFASSUNG
Ultrastructural localization of insulin in B cells of guinea pig pancreas was accomplished after osmium fixation with an immunoperoxidase procedure that utilized 4-chloro-1-naphthol (CN) in the substrate solution. The principal features of this protocol were: a) osmium tetroxide postfixation; b) omission of hydrogen peroxide "etching"; c) use of CN instead of diaminobenzidine in the substrate solution; d) elimination of osmium tetroxide after the substrate reaction; e) uranyl acetate and lead citrate counterstaining. This procedure produces intense specific staining with low background using highly dilute antiserum, and appears to be useful for postembedding immunoperoxidase staining of a variety of peptide antigens in osmium-fixed tissue.