- Immobilization of protein molecules on liposomes. Anchorage by artificially bound unsaturated hydrocarbon tails.
Immobilization of protein molecules on liposomes. Anchorage by artificially bound unsaturated hydrocarbon tails.
Biochemical pharmacology (1983-04-01)
V S Goldmacher
PMID6189496
ZUSAMMENFASSUNG
A method for the immobilization of trypsin, a hydrophilic nonmembrane protein, on a liposomal surface has been developed. The technique consists of covalent coupling of linoleoyl residues to the protein globules and consequent binding of linoleoyl trypsin to liposomes by a detergent dilution method. The immobilized protein preserved its biological functions: specific esterolytic catalytic activity and ability to bind to a macromolecular trypsin protein inhibitor. Liposomes carrying immobilized trypsin were able to sequester glucose with the same efficiency as liposomes without trypsin.
MATERIALIEN