Merck
  • Influence of vehicles on penetration through human epidermis of benzyl alcohol, isoeugenol and methyl isoeugenol.
Influence of vehicles on penetration through human epidermis of benzyl alcohol, isoeugenol and methyl isoeugenol.
The Journal of dermatology (1983-06-01)
Y Jimbo, M Ishihara, H Osamura, M Takano, M Ohara
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylisoeugenol, ≥98%, FG
Sigma-Aldrich
1,2-Dimethoxy-4-Propenylbenzol, 99%