Merck
  • Interaction between neurotoxicities induced by organophosphorus and long-chain hexacarbon compounds.
Interaction between neurotoxicities induced by organophosphorus and long-chain hexacarbon compounds.
Neurotoxicology (1983-01-01)
M B Abou-Donia
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,5-Hexandiol, 99% (mixture of isomers)