Merck
  • 19F nuclear magnetic resonance observations of aldehyde dismutation catalyzed by horse liver alcohol dehydrogenase.
19F nuclear magnetic resonance observations of aldehyde dismutation catalyzed by horse liver alcohol dehydrogenase.
Archives of biochemistry and biophysics (1982-08-01)
D C Anderson, F W Dahlquist
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-(Trifluormethyl)benzaldehyd, 98%