Merck
  • Replacement of ethanolamine phosphate by 3-aminopropylphosphonate in the phospholipids of Tetrahymena.
Replacement of ethanolamine phosphate by 3-aminopropylphosphonate in the phospholipids of Tetrahymena.
Archives of biochemistry and biophysics (1981-02-01)
J D Smith, D A Giegel
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Aminopropylphosphonsäure, 98%