Merck
Effects of antilipolytic agents on peroxisomal beta-oxidation of fatty acids in rat liver.
Biochemical pharmacology (1983-06-01)
T Locci-Cubeddu, E Bergamini
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,5-Dimethyl-pyrazol, 99%
Sigma-Aldrich
3,5-Dimethyl-pyrazol, Wacker Chemie AG, ≥99.0% (GC)