Merck
A conductometric method for the assay of amidase and peptidase activities.
Analytical biochemistry (1982-02-01)
C R Hill, G Tomalin
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Leucinamid -hydrochlorid, 99%