Merck
  • Effect of some new antibilharzial drugs on oxygen consumption of mouse diaphragm and liver homogenates in experimental bilharziasis.
Effect of some new antibilharzial drugs on oxygen consumption of mouse diaphragm and liver homogenates in experimental bilharziasis.
Journal of the Egyptian Society of Parasitology (1982-06-01)
S M Amin, N A Hammouda, Y A Habib, A R Mikhail, L A Hameid
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1H-1,2,4-Triazol-3-thiol, 97%