Merck
The reaction of poly-L-ornithine with 1-halo-2,4-dinitrobenzenes.
The International journal of biochemistry (1981-01-01)
R C Parker, R P Gural, D S Kristol
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Brom-2,4-dinitrobenzol, 97%