Merck
  • Water-soluble siomycin-A derivatives. Preparation, chemical structures and biological properties of half-esters of the peptide antibiotic.
Water-soluble siomycin-A derivatives. Preparation, chemical structures and biological properties of half-esters of the peptide antibiotic.
The Journal of antibiotics (1981-07-01)
K Tokura, K Hayashi, K Okabe, K Tori, Y Yoshimura, M Mayama, S Matsuura, H Otsuka
ZUSAMMENFASSUNG
The peptide antibiotic siomycin-A was transformed into half-esters with dicarboxylic acids with the intention of making siomycin-A soluble in water. Sodium salts of the half-esters were also prepared. Some of the salts showed antibacterial activities comparable to siomycin-A against Gram-positive bacteria in vitro and exhibited better therapeutic effects in infected mice than siomycin-A. The chemical structures of siomycin-A hemiadipate-II and -III were elucidated by comparing their 13C and 1H NMR spectra with those of siomycin-A. Their physicochemical properties are described.