Merck
Lactate dehydrogenase isoenzymes in mammalian cells exposed to isophenphos.
Biomedical and environmental sciences : BES (1995-03-01)
M A Rodrigues, M J Fernandes, M D Angelo
ZUSAMMENFASSUNG
The cytotoxicity of isophenphos, an organophosphorus insecticide that has the potential to cause delayed polyneuropathy, was evaluated in GBK and V79 cells. A 72 h time course following isophenphos exposure indicated a dose-dependent growth inhibition as determined by cell counts. The administration of isophenphos (20 g/ml) to GBK cells cultured at high densities indicated a decrease in the activities of LDH isoenzymes. Analysis of V79 cells revealed a decrease of LDH3, the only LDH isoenzyme detected in these cells.