Merck
Enzymatic production of cephalexin.
Enzyme and microbial technology (1994-08-01)
N K Maladkar
ZUSAMMENFASSUNG
Enzymatic production of cephalexin using immobilized penicillin G acylase was studied. The factors that affect the conversion of 7-amino-3-deacetoxycephalosporanic acid (7-ADCA) to cephalexin, such as pH; temperature; concentrations of 7-ADCA, D(-)-phenylglycine methyl ester (PGME), and immobilized penicillin G acylase (IMPGA); time; and molar ratios of 7-ADCA and PGME, were investigated. Under optimized conditions, 85% conversion of 7-ADCA to cephalexin was achieved. The IMPGA was used for 10 cycles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
7-Aminodesacetoxycephalosporansäure