Merck
  • Rapid determination of dabsylated hydroxyproline from cultured cells by reversed-phase high-performance liquid chromatography.
Rapid determination of dabsylated hydroxyproline from cultured cells by reversed-phase high-performance liquid chromatography.
Journal of chromatography. B, Biomedical applications (1995-02-17)
K Sormiachi, M Ikeda, K Akimoto, A Niwa
ZUSAMMENFASSUNG
A high-performance liquid chromatographic method was modified for the determination of hydroxyproline in cultured cells derived from rat liver. First, the primary amino group in the cell hydrolysate was blocked with o-phthalaldehyde, then the secondary amino group was derivatized with 4-dimethylaminoazobenzene-4'-sulfonyl chloride (dabsyl chloride). The dabsylated sample was treated with ethyl acetate to obtain a simple chromatographic elution profile of the cell hydrolysate. Dabsylhydroxyproline and proline were separated from other compounds by high-performance liquid chromatography in the gradient elution mode, and eluted at 4.71 and 8.00 min, respectively. The method was applied to the determination of hydroxyproline contained in cultured cells, the result being 25.4 +/- 3.6 pmol/microgram.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-(4-Dimethylaminophenylazo)-benzolsulfonylchlorid, ≥97.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.