Merck
Chemical modification of ovine prolactin with N-acetylimidazole.
International journal of peptide and protein research (1993-07-01)
G D Cymes, M M Iglesias, C Wolfenstein-Todel
ZUSAMMENFASSUNG
Reaction of ovine prolactin (oPRL) with a 150-fold molar excess of N-acetylimidazole over protein content resulted in the modification of 2.5 tyrosine residues and 1.2 lysine residues. Acetylation greatly decreased the in vitro binding capacity to lactogenic sites. This binding capacity was partially restored by ammonium bicarbonate treatment, which removes O-acetyl groups from tyrosine residues but not N-acetyl groups from lysine residues. The modification extent of the tyrosine residues was determined. The results suggest that acetylation of tyrosine 44 or of tyrosine 96 is likely to be responsible for the decrease in binding activity of acetylated oPRL, and that one of these residues may play a role in the interaction of oPRL with lactogenic receptors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Acetylimidazol, 98%