Merck
  • Identification of the sites of modification on bovine carbonic anhydrase II (BCA II) by the salt-bridge reagent cyanogen, C2N2.
Identification of the sites of modification on bovine carbonic anhydrase II (BCA II) by the salt-bridge reagent cyanogen, C2N2.
Biochimica et biophysica acta (1993-01-15)
G Ghenbot, T Emge, R A Day
ZUSAMMENFASSUNG
The hydrolase activities of bovine carbonic anhydrase B (BCA II carbonate hydrolyase, EC 4.2.1.1) were modified by cyanogen (C2N2, N identical to C-C identical to N, ethanedinitrile) with decreases in Vmax of as much as 99%. This was not accompanied by a reduction in hydrolyase activity. These changes were not reversed at lower pH values but the enzymatic activity was restored by incubation at pH 10. 14C-labeled glycine ethyl ester ([14C]GEE) specifically and covalently bound to the cyanogen-treated BCA II, as verified by HPLC and 14C monitoring. It was shown that sites of cyanogen-introduced modifications in BCA II could be effectively labeled and identified by incubation with the nucleophile [14C]GEE. Three radiolabeled tryptic peptides from BCA II arising from a labeling process designed to study cyanogen-induced modifications leading to nucleophile labile covalent bonds have been isolated. The residues identified by [14C]GEE labeling were Asp-34, Glu-117 and Asp-152. Three moieties attached to the omega-carboxyls by C2N2 were tentatively identified by molecular modeling; they were Arg-111, His-107 and/or His-119 and Ser-216, respectively. The use of C2N2 afforded a means to compare the salt bridges in two species and showed that two of three were not conserved.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycinethylester -hydrochlorid, 99%